
Quinidine gluconate

Synonym(s):
Quinidine gluconate salt
Empirical Formula (Hill Notation):
C20H24N2O2 · C6H12O7
CAS Number:
Molecular Weight:
520.57
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2