
Monoclonal Anti-MDM2 antibody produced in mouse

Synonym(s):
Anti-MGC71221, Anti-Mdm2, transformed 3T3 cell double minute 2, p53 binding protein (mouse), Anti-hdm2
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing