
Methylpyrrolidone

Synonym(s):
1-Methyl-2-pyrrolidinone, 1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP
Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing