
Methylophiopogonanone A

Synonym(s):
(3R)-3-(1,3-Benzodioxol-5-ylmethyl)-2,3-dihydro-5,7-dihydroxy-6,8-dimethyl-4H-1-benzopyran-4-one
Empirical Formula (Hill Notation):
C19H18O6
CAS Number:
Molecular Weight:
342.34
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing