
Methyl caproate

Synonym(s):
Methyl hexanoate, Caproic acid methyl ester, Hexanoic acid methyl ester, Methyl caproate
Linear Formula:
CH3(CH2)4COOCH3
CAS Number:
Molecular Weight:
130.18
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing