
Idarubicin hydrochloride

Synonym(s):
(7S-cis)-9-Acetyl-7-[(3-amino-2,3,6-trideoxy-α-L-lyxo-hexopyranosyl)oxy]-7,8,9,10-tetrahydro-6,9,11-trihydroxy-5,12-naphthacenedione, 4-Demethoxydaunorubicin hydrochloride, DMDR, IMI-30, Idamycin
Empirical Formula (Hill Notation):
C26H27NO9 · HCl
CAS Number:
Molecular Weight:
533.95
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing