![Grubbs Catalyst® M207 Umicore](/deepweb/assets/sigmaaldrich/product/structures/347/315/47b10e48-a567-4d6f-aecf-0ee8785657a5/640/47b10e48-a567-4d6f-aecf-0ee8785657a5.png)
Grubbs Catalyst® M207
![Grubbs Catalyst® M207 Umicore](/deepweb/assets/sigmaaldrich/product/structures/347/315/47b10e48-a567-4d6f-aecf-0ee8785657a5/640/47b10e48-a567-4d6f-aecf-0ee8785657a5.png)
Synonym(s):
Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene](3-methyl-2-butenylidene) (tricyclohexylphosphine)ruthenium(II), Isopentenylidene(1,3-dimesitylimidazolidin-2-ylidene) (tricyclohexylphosphine)ruthenium(II) dichloride, [SIMes]dichloro(3-methyl-2-butenylidene)(tricyclohexylphosphine)Ru(II)
Empirical Formula (Hill Notation):
C44H67Cl2N2PRu
CAS Number:
Molecular Weight:
826.97
Products
Product Category
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing