
Glycopyrrolate Related Compound I

Synonym(s):
(RS)-3-[(SR)-2-(4-Chlorophenyl)-2-cyclopentyl-2-hydroxyacetoxy]-1,1-dimethylpyrrolidin-1-ium bromide
Empirical Formula (Hill Notation):
C19H27BrClNO3
Molecular Weight:
432.78
Products
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing