
Ginkgolic acid C13:0

Synonym(s):
2-Hydroxy-6-tridecylbenzoic acid, 6-Tridecylsalicylic acid, Ginkgoneolic acid
Empirical Formula (Hill Notation):
C20H32O3
CAS Number:
Molecular Weight:
320.47
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing