
Gentiopicroside

Synonym(s):
(3S,4R)-4-ethenyl-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4,6-dihydro-3H-pyrano[3,4-c]pyran-8-one, (5R,6S)-5-Ethenyl-6-(-D-glucopyranosyloxy)-5,6-dihydro-1H,3H-Pyrano[3,4-c]pyran-1-one, (5R-trans)-5-ethenyl-6-(-D-glucopyranosyloxy)-5,6-dihydro-1H,3H-pyrano[3,4-c]pyran-1-one, Gentiopicrin, NSC606402
Empirical Formula (Hill Notation):
C16H20O9
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing