
Fura 2 pentapotassium salt

Empirical Formula (Hill Notation):
C29H22K5N3O14
CAS Number:
Molecular Weight:
831.99
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing