
Fosinopril Related Compound G

Synonym(s):
2-[Hydroxy(4-phenylbutyl)phosphinyl]acetic acid sodium salt, (4-Phenylbutyl)phosphinyl acetic acid sodium salt
Empirical Formula (Hill Notation):
C12H15Na2O4P
CAS Number:
Molecular Weight:
300.20
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing