
EX-CELL® ACF CHO Medium

Synonym(s):
CHO Medium
Products
Product Category
Brand
Physical Form
HEPES (mM)
L-Glutamine
Sodium Bicarbonate
Phenol Red
Quality Segment
Sterilization Method
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing