
Everolimus

Synonym(s):
Everolimus, 42-O-(2-Hydroxyethyl)rapamycin
Empirical Formula (Hill Notation):
C53H83NO14
CAS Number:
Molecular Weight:
958.22
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3