Skip to Content
Merck
Eicosane analytical standard

Eicosane

Eicosane analytical standard
Linear Formula:
CH3(CH2)18CH3
CAS Number:
Molecular Weight:
282.55
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique