
Echinacoside

Empirical Formula (Hill Notation):
C35H46O20
CAS Number:
Molecular Weight:
786.73
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4