
D-myo-Inositol 1,3,4,5-tetrakis(phosphate) potassium salt

Synonym(s):
IP4
Empirical Formula (Hill Notation):
C6H8O18P4K8
CAS Number:
Molecular Weight:
804.80
Products
Brand
Formula Weight
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing