
Desmethyl-formamido-pirimicarb

Synonym(s):
5,6-Dimethyl-2-(N-formylmethylamino)-4-pyrimidinyl dimethylcarbamate
Empirical Formula (Hill Notation):
C11H16N4O3
CAS Number:
Molecular Weight:
252.27
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2