
Deblocking Reagent 3% Dichloroacetic Acid in Toluene

Synonym(s):
DCA Deblock
Empirical Formula (Hill Notation):
C2H2Cl2O2
CAS Number:
Molecular Weight:
128.94
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing