
Clindamycin phosphate for system suitability

Synonym(s):
Clindamycin 2-phosphate, Clindamycin 2-dihydrogen phosphate
Empirical Formula (Hill Notation):
C18H34ClN2O8PS
CAS Number:
Molecular Weight:
504.96
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing