![β-Nicotinamide adenine dinucleotide phosphate hydrate](/deepweb/assets/sigmaaldrich/product/structures/246/556/ce856d88-2ba7-4cc6-a245-7d37c1a31bf7/640/ce856d88-2ba7-4cc6-a245-7d37c1a31bf7.png)
β-Nicotinamide adenine dinucleotide phosphate hydrate
![β-Nicotinamide adenine dinucleotide phosphate hydrate](/deepweb/assets/sigmaaldrich/product/structures/246/556/ce856d88-2ba7-4cc6-a245-7d37c1a31bf7/640/ce856d88-2ba7-4cc6-a245-7d37c1a31bf7.png)
Synonym(s):
β-NADP, Coenzyme II, NADP, TPN, Triphosphopyridine nucleotide
Empirical Formula (Hill Notation):
C21H28N7O17P3 · xH2O
CAS Number:
Molecular Weight:
743.41 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing