
Bicalutamide impurity D

Synonym(s):
4-Amino-2-(trifluoromethyl)benzonitrile, 4-Cyano-3-trifluoromethylaniline
Linear Formula:
NCC6H3(CF3)NH2
CAS Number:
Molecular Weight:
186.13
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing