
Astragaloside I

Synonym(s):
(3β,6α,16β,20R,24S)-3-[(2,3-Di-O-acetyl-β-D-xylopyranosyl)oxy]-20,24-epoxy-16,25-dihydroxy-9,19-cyclolanostan-6-yl β-D-glucopyranoside, Astrasieversianin IV, Cyclosieversioside B
Empirical Formula (Hill Notation):
C45H72O16
CAS Number:
Molecular Weight:
869.04
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1