
Aluminium nitrate nonahydrate

Synonym(s):
Aluminium nitrate nonahydrate
Linear Formula:
Al(NO3)3 · 9 H2O
CAS Number:
Molecular Weight:
375.13
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing