
Alfuzosin for peak identification

Synonym(s):
Alfuzosin hydrochloride, N-(3-((4-Amino-6,7-dimethoxy-2-quinazolinyl)methylamino)propyl)tetrahydro-2-furancarboxamide hydrochloride, Uroxatral hydrochloride
Empirical Formula (Hill Notation):
C19H27N5O4 · HCl
CAS Number:
Molecular Weight:
425.91
Products
Formula Weight
Gene Alias
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing