
Aclonifen

Synonym(s):
2-Chloro-6-nitro-3-phenoxyaniline
Empirical Formula (Hill Notation):
C12H9ClN2O3
CAS Number:
Molecular Weight:
264.66
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2