
4-Bromobenzaldehyde

Synonym(s):
4-Bromobenzaldehyde
Linear Formula:
4-(Br)C6H4CHO
CAS Number:
Molecular Weight:
185.02
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing