
2-Bromopropane

Synonym(s):
Isopropyl bromide, 2-Bromopropane
Linear Formula:
(CH3)2CHBr
CAS Number:
Molecular Weight:
122.99
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing