Skip to Content
Merck
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate for chiral derivatization, LiChropur™, ≥98.0% (HPLC)

2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate

2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate for chiral derivatization, LiChropur™, ≥98.0% (HPLC)
Synonym(s):
GITC
Empirical Formula (Hill Notation):
C15H19NO9S
CAS Number:
Molecular Weight:
389.38

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing