
1-Propanethiol

Synonym(s):
n-Propylmercaptan, Mercaptan C3, 1-Propanethiol
Linear Formula:
CH3CH2CH2SH
CAS Number:
Molecular Weight:
76.16
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing