Skip to Content
Merck
All Photos(1)

Key Documents

SML3216

Sigma-Aldrich

Melarsoprol

≥98% (HPLC)

Synonym(s):

(2-(4-(4,6-diamino-1,3,5-triazin-2-ylamino)phenyl)-1,3,2-dithiarsolan-4-yl)methanol, 2-(4-melamin-2-ylphenyl)-4-hydroxymethyl-1,3-dithia-2-arsolane, 2-p-(4,6-diamino-s-triazin-2-ylamino)phenyl-4-hydroxymethyl-1,3,2-dithiarsoline, Mel B, Melarsaprol, Melarsen B, RP 3854, p-[(4,6-diamino-s-triazin-2-yl)amino]dithiobenzenearsonous acid 3-hydroxypropylene ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15AsN6OS2
CAS Number:
Molecular Weight:
398.34
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=NC(N)=NC(NC2=CC=C(C=C2)[As]3SCC(S3)CO)=N1

InChI

1S/C12H15AsN6OS2/c14-10-17-11(15)19-12(18-10)16-8-3-1-7(2-4-8)13-21-6-9(5-20)22-13/h1-4,9,20H,5-6H2,(H5,14,15,16,17,18,19)

InChI key

JCYZMTMYPZHVBF-UHFFFAOYSA-N

Biochem/physiol Actions

An antiprotozoal (Trypanosoma) drug used in treatment of trypanosomiasis (Chagas disease and West African sleeping sickness);
Melarsoprol is an antiprotozoal (Trypanosoma) drug used in treatment of trypanosomiasis (Chagas disease and West African sleeping sickness). The drug mechanism of action is not well characterized. Melarsoprol is still the only effective drug for chemotherapy of second stage of T. b. rhodesiense sleeping sickness. As other arsenic-containing compounds melarsoprol causes cell apoptosis of myeloma and other cancer cell lines.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service