SAB2702341
Monoclonal Anti-ARID1A antibody produced in mouse
clone GT3611, affinity isolated antibody
Synonym(s):
B120, BAF250, BAF250a, BM029
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
GT3611, monoclonal
form
buffered aqueous solution
mol wt
242kDa
species reactivity
human, mouse
concentration
2.61mg/mL
technique(s)
immunofluorescence: 1:100-1:1,000
immunohistochemistry: 1:100-1:1,000
western blot: 1:500-1:3,000
isotype
IgG2b
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... ARID1A(8289)
Immunogen
Application
Biochem/physiol Actions
Features and Benefits
Other Notes
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service