Skip to Content
Merck
All Photos(1)

Key Documents

ABS553

Sigma-Aldrich

Anti-phospho KDR/FLK-1/VEGFR2 Antibody (Tyr1059)

from rabbit, purified by affinity chromatography

Synonym(s):

Vascular endothelial growth factor receptor 2, VEGFR-2, Fetal liver kinase 1, FLK-1, Kinase insert domain receptor, KDR, Protein-tyrosine kinase receptor flk-1, CD309

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

sheep (based on 100% sequence homology), chicken (based on 100% sequence homology), bovine (based on 100% sequence homology), canine (based on 100% sequence homology), zebrafish (based on 100% sequence homology)

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr1059)

Gene Information

human ... KDR(3791)

General description

VEGFR2, also known as Fetal liver kinase 1 (FLK-1), Kinase insert domain receptor (KDR), Protein-tyrosine kinase receptor flk-1, or CD309, and encoded by the gene KDR/FLK1/VEGFR2, is a tyrosine-protein kinase that acts as a cell-surface receptor for VEGFA, VEGFC and VEGFD. VEGFR2 plays an essential role in the regulation of angiogenesis, vascular development, vascular permeability, and embryonic hematopoiesis. It also promotes proliferation, survival, migration and differentiation of endothelial cells and influences the reorganization of the actin cytoskeleton. Binding of vascular growth factors to isoform 1 leads to the activation of several signaling cascades including the inositol 1,4,5-trisphosphate pathway and the activation of protein kinase C, and the MAP kinase, AKT and FAK pathways. VEGFR2 is also required for VEGFA-mediated induction of NOS2 and NOS3, leading to the production of the signaling molecule nitric oxide (NO) by endothelial cells. VEGFR2 isoform 1 is cell membrane bound; isoform 2, 3 are secreted.

Immunogen

Epitope: Cytoplasmic domain
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human KDR/FLK-1/VEGFR2 phosphorylated at Tyr1059.

Application

Peptide Inhibition Analysis: A 1:500 dilution from a representative lot detected KDR/FLK-1/VEGFR2 in CKR-PAE cell lysates treated with Macrophage Colony Stimulating Factor when antibody was preincubated with or without non-phospho peptide. This antibody did not detect KDR/FLK-1/VEGFR2 in CKR-PAE cell lysates treated with Macrophage Colony Stimulating Factor when antibody was preincubated with corresponding phospho peptide.
Research Category
Signaling
Research Sub Category
Cell Cycle, DNA Replication & Repair
This Anti-phospho KDR/FLK-1/VEGFR2 Antibody (Tyr1059) is validated for use in western blotting & peptide inhibition assay for the detection of phospho KDR/FLK-1/VEGFR2 (Tyr1059).

Quality

Evaluated by Western Blotting in untreated and VEGF treated HUVEC cell lysates.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected KDR/FLK-1/VEGFR2 in VEGF treated HUVEC cell lysate.

Target description

~230 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donghui Liu et al.
Scientific reports, 6, 34649-34649 (2016-10-04)
Sodium butyrate (NaBu), a form of short-chain fatty acid (SCFA), acts classically as a potent anti-angiogenic agent in tumour angiogenesis models, some authors demonstrated that low concentrations of NaBu may contribute to healing of tendon-bone injury in part at least
Syndecan-2 selectively regulates VEGF-induced vascular permeability.
Corti, et al.
Nature cardiovascular research, 1, 518-528 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service