Skip to Content
Merck
All Photos(1)

Key Documents

341286

Sigma-Aldrich

Anti-Farnesyl Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity (predicted by homology)

all

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Farnesyl, rabbit polyclonal, recognizes farnesyl in a wide variety of species. Also recognizes KLH. It is validated for use in ELISA and immunofluorescence.
Purified rabbit polyclonal antibody. Recognizes farnesyl.
Recognizes farnesyl. Also recognizes KLH.

Immunogen

farnesyl cysteine conjugated to KLH

Application

ELISA (1:1000)

Immunofluorescence (1:50)

Immunoblotting (not recommended; see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Baron, R., et al. 2000. Proc. Natl. Acad. Sci. USA97, 11626.
Lin, H.P., et al. 1999. J. Gen. Virol.80, 91.
Aspbury, R.A., et al. 1998. Biochim Biophys Acta1392, 265.
Gromov, P.S., et al. 1996. Electrophoresis17, 1728.
Zhang, F.L. & Casey, P.J. 1996. Annu. Rev. Biochem.65, 241.
Recognizes proteins that have been isoprenylated by farnesyltransferase of geranylgeranyltransferase at the C-A-A-X motif. Also recognizes KLH. Not suitable for immunoblotting due to the hydrophobicity of the isoprenylated motif. Variables associated with assay conditions will dictate the optimal working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimo Bonora et al.
Cell stem cell, 31(3), 359-377 (2024-03-09)
Mitochondrial fatty acid oxidation (FAO) is essential for hematopoietic stem cell (HSC) self-renewal; however, the mechanism by which mitochondrial metabolism controls HSC fate remains unknown. Here, we show that within the hematopoietic lineage, HSCs have the largest mitochondrial NADPH pools

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service