Skip to Content
Merck
All Photos(2)

Key Documents

700069P

Avanti

lathosterol

Avanti Research - A Croda Brand

Synonym(s):

5α-cholest-7-en-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (700069P-10mg)
pkg of 1 × 5 mg (700069P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC1CC2C(C3CCC4(C(CCC4C(CCCC(C)C)C)C3=CC2)C)(CC1)C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h10,18-21,23-25,28H,6-9,11-17H2,1-5H3

InChI key

IZVFFXVYBHFIHY-UHFFFAOYSA-N

General description

Lathosterol is a cholesterol synthesis precursor.[1]

Application

Lathosterol has been used as a standard for the estimation of the concentrations of cholesterol and plant sterols.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700069P-10mg)
5 mL Amber Glass Screw Cap Vial (700069P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid Analyses of Four Types of Fish From Manitoba Lakes
Moghadasian MH, et al.
EC nutrition, 1(2), 41-48 (2015)
Lathosterol and other non-cholesterol sterols during treatment of hypercholesterolaemia with beta-glucan-rich oat bran
UusitupaMIJ, et al.
European Journal of Clinical Nutrition, 51(9), 607-607 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service