SLW5808/3
Pyrex® Filter holder for 25 mm membranes
support base with sintered glass disc only
Synonym(s):
Pyrex, SciLabware 5808/3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
description
fits filter 25 mm
manufacturer/tradename
5808/3
SciLabware
pore size
grade 3 porosity (sintered glass disc)
Looking for similar products? Visit Product Comparison Guide
General description
- With porosity grade 3 sintered glass disc to support 25mm diameter membranes
- Liquids only in contact with chemically resistant borosilicate glass
- Fully autoclavable
Comprises three components which should be ordered separately
- Funnel 15ml capacity
- Support base with sintered glass disc
- Metal retaining clip
Features and Benefits
• With porosity grade 3 sintered glass disc to support 25 mm diameter membranes• Liquids only in contact with chemically resistant borosilicate glass• Comprises three components which should be ordered separately: - Graduated 15 ml capacity funnel - Support base with sintered glass disc - Metal retaining clip• Fully autoclavable
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service