Skip to Content
Merck
All Photos(1)

Key Documents

455393

Sigma-Aldrich

4-Fluoro-2-(trifluoromethyl)benzoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CF3)COCl
CAS Number:
Molecular Weight:
226.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.468 (lit.)

bp

157 °C (lit.)

density

1.495 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1ccc(C(Cl)=O)c(c1)C(F)(F)F

InChI

1S/C8H3ClF4O/c9-7(14)5-2-1-4(10)3-6(5)8(11,12)13/h1-3H

InChI key

OEYHURRIOWWRMD-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert T Jacobs et al.
Future medicinal chemistry, 3(10), 1259-1278 (2011-08-24)
Human African trypanosomiasis, caused by the kinetoplastid parasite Trypanosoma brucei, affects thousands of people across sub-Saharan Africa, and is fatal if left untreated. Treatment options for this disease, particularly stage 2 disease, which occurs after parasites have infected brain tissue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service