HPLC Analysis of 1,1′-Binaphthyl-2,2′-diyl Hydrogenphosphate Enantiomers on Astec® (R,R) P-CAP™
![HPLC Analysis of 1,1′-Binaphthyl-2,2′-diyl Hydrogenphosphate Enantiomers on Astec® (R,R) P-CAP™ application for HPLC](/deepweb/assets/sigmaaldrich/product/images/302/939/bca4adca-5fed-4357-83db-29b4ee3eefda/800/bca4adca-5fed-4357-83db-29b4ee3eefda.jpg)
CONDITIONS
column
Astec (R,R) P-CAP, 25 cm x 4.6 mm I.D., 5 μm particles (31024AST)
mobile phase
20 mM ammonium acetate in: (A) acetonitrile; (B) methanol; (70:30, A:B)
flow rate
0.9 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
5 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
P-CAP is a trademark of Sigma-Aldrich Co. LLC