Merck
  • Automated measurement and calibration of reactive volatile halogenated organic compounds in the atmosphere.
Automated measurement and calibration of reactive volatile halogenated organic compounds in the atmosphere.
The Analyst (2004-06-24)
David J Wevill, Lucy J Carpenter
ABSTRACT
The automated calibration and analysis of very low mixing ratios of the reactive volatile organic halocarbons CH(3)I, CHCl(3), C(2)H(5)I, 2-C(3)H(7)I, CH(2)Br(2), CH(2)ClI, CHBr(2)Cl, 1-C(3)H(7)I, CH(2)BrI, CHBr(3) and CH(2)I(2) for long term atmospheric field measurements are described. Analytes were pre-concentrated from 3 l of air onto an adsorbent trap cooled to -10 [degree]C using Peltier plates, and rapidly transferred to a gas chromatograph (GC) by resistive heating. A two stage Carboxen 1016/Carbotrap C adsorbent trap allowed good analyte recovery and rapid desorption without the need for post-desorption cryofocussing. Halocarbons were detected using a mass spectrometer (MS) in selective ion mode. Detection limits were between 0.02 and 0.12 pptv (parts per trillion by volume) for approximately hourly samples of CHCl(3), CH(3)I, C(2)H(5)I, 1-C(3)H(7)I, 2-C(3)H(7)I, CH(2)ClI, CH(2)Br(2), CHBr(2)Cl, CH(2)BrI, CHBr(3) and CH(2)I(2) with a precision of 3-8%. A novel calibration system was constructed which utilised fixed volume (50 [micro sign]l) injections of the output of thermostatted permeation tubes into a stream of nitrogen gas in order to dilute parts per million by volume (ppmv) mixing ratios into pptv. The calibration was completely automated, allowing multi-point calibrations during routine operation. The overall accuracy of the measurements is estimated to be +/-15%. The instrument was used continuously for automated atmospheric measurements during a 4-month research cruise from Germany to Antarctica, and a 6 week field campaign at Mace Head, Ireland. The results for CHCl(3) during the latter campaign were within 13% of measurements made by a GC-MS operating continuously at the site within the long term Advanced Global Atmospherics Gases Experiment.
Product Description
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
Carbotrap® 349, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Supelco
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® B, 20-40 mesh, bottle of 10 g
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Supelco
Carbotrap® 300, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Supelco
Carbotrap® 300, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, preconditioned, pkg of 10 ea
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 10 g
Supelco
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 1/4 in. × 5 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® X, 20-40 mesh, bottle of 10 g
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm