Merck
Leukotriene B
Scientific reports (2017-12-17)
Morgana K B Prado, Gisele A Locachevic, Karina F Zoccal, Francisco W G Paula-Silva, Caroline Fontanari, Joseane C Ferreira, Priscilla A T Pereira, Luiz G Gardinassi, Simone G Ramos, Carlos A Sorgi, Ana Lúcia C Darini, Lúcia H Faccioli
ABSTRACT
Leukotriene B
Sigma-Aldrich
BLT-1, ≥98% (HPLC)