Merck
  • Nuclear receptor SHP, a death receptor that targets mitochondria, induces apoptosis and inhibits tumor growth.
Nuclear receptor SHP, a death receptor that targets mitochondria, induces apoptosis and inhibits tumor growth.
Molecular and cellular biology (2010-01-13)
Yuxia Zhang, Jamie Soto, Kyungtae Park, Gunda Viswanath, Scott Kuwada, E Dale Abel, Li Wang
ABSTRACT
Small heterodimer partner (SHP) is an epigenetically regulated nuclear transcriptional repressor that suppresses the development of liver cancer by inhibiting cellular growth. Here we report a novel cytoplasmic function of SHP through its regulation of mitochondrial activity. SHP is a pivotal cell death receptor that targets mitochondria, where it binds with Bcl-2, disrupts Bcl-2/Bid interaction, and induces cytochrome c release. The apoptosis inducer AHPN {retinoid 6-[3-(1-adamantyl)-4-hydroxyphenyl]-2-naphthalene carboxylic acid} acts by regulating SHP gene expression and promotes the translocation of SHP from the nucleus to the mitochondria. Induction of apoptosis by SHP activation inhibits peritoneal pancreatic tumor growth. Our findings provide for the first time a mechanism by which SHP regulates cell survival, namely, by controlling mitochondrial function via modulating the activity of Bcl-2 through AHPN-mediated or AHPN-independent action. Thus, SHP regulates a mechanism by which apoptotic signals can mediate local control of mitochondrial function and apoptosis, which in turn may limit tumorigenesis.
Product Description
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)