Merck
  • Selective Removal of FG Repeat Domains from the Nuclear Pore Complex by Enterovirus 2A(pro).
Selective Removal of FG Repeat Domains from the Nuclear Pore Complex by Enterovirus 2A(pro).
Journal of virology (2015-08-28)
Nogi Park, Nicholas J Schweers, Kurt E Gustin
ABSTRACT
Enteroviruses proteolyze nuclear pore complex (NPC) proteins (Nups) during infection, leading to disruption of host nuclear transport pathways and alterations in nuclear permeability. To better understand how enteroviruses exert these effects on nuclear transport, the mechanisms and consequences of Nup98 proteolysis were examined. The results indicate that Nup98 is rapidly targeted for degradation following enterovirus infection and that this is mediated by the enterovirus 2A protease (2A(pro)). Incubation of bacterially expressed or in vitro-translated Nup98 with 2A(pro) results in proteolytic cleavage at multiple sites in vitro, indicating that 2A(pro) cleaves Nup98 directly. Site-directed mutagenesis of putative cleavage sites identified Gly374 and Gly552 as the sites of 2A(pro) proteolysis in Nup98 in vitro and in infected cells. Indirect immunofluorescence assays using an antibody that recognizes the N terminus of Nup98 revealed that proteolysis releases the N-terminal FG-rich region from the NPC. In contrast, similar analyses using an antibody to the C terminus indicated that this region is retained at the nuclear rim. Nup88, a core NPC component that serves as a docking site for Nup98, also remains at the NPC in infected cells. These findings support a model whereby the selective removal of Nup FG repeat domains leads to increased NPC permeability and inhibition of certain transport pathways, while retention of structural domains maintains the overall NPC structure and leaves other transport pathways unaffected. Enteroviruses are dependent upon host nuclear RNA binding proteins for efficient replication. This study examines the mechanisms responsible for alterations in nuclear transport in enterovirus-infected cells that lead to the cytoplasmic accumulation of these proteins. The results demonstrate that the enterovirus 2A protease directly cleaves the nuclear pore complex (NPC) protein, Nup98, at amino acid positions G374 and G552 both in vitro and in infected cells. Cleavage at these positions results in the selective removal of the FG-containing N terminus of Nup98 from the NPC, while the C terminus remains associated. Nup88, a core component of the NPC that serves as a docking site for the C terminus of Nup98, remains associated with the NPC in infected cells. These findings help to explain the alterations in permeability and nuclear transport in enterovirus-infected cells and how NPCs remain functional for certain trafficking pathways despite significant alterations to their compositions.
Sigma-Aldrich
Anti-Nup98 antibody, Rat monoclonal, clone 2H10, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)