Merck
  • EBV-miR-BART10-3p facilitates epithelial-mesenchymal transition and promotes metastasis of nasopharyngeal carcinoma by targeting BTRC.
EBV-miR-BART10-3p facilitates epithelial-mesenchymal transition and promotes metastasis of nasopharyngeal carcinoma by targeting BTRC.
Oncotarget (2015-10-27)
Qijia Yan, Zhaoyang Zeng, Zhaojian Gong, Wenling Zhang, Xiayu Li, Baoyu He, Yali Song, Qiao Li, Yong Zeng, Qianjin Liao, Pan Chen, Lei Shi, Songqing Fan, Bo Xiang, Jian Ma, Ming Zhou, Xiaoling Li, Jianbo Yang, Wei Xiong, Guiyuan Li
ABSTRACT
Epstein-Barr virus (EBV) infection is closely associated with tumorigenesis and development of nasopharyngeal carcinoma (NPC), but the underlying molecular mechanisms remain poorly understood. It has been recently reported that EBV encodes 44 mature miRNAs, some of which were found to promote tumor development by targeting virus-infected host genes or self-viral genes. However, few targets of EBV encoded-miRNAs that are related to NPC development have been identified to date. In this study, we revealed that in NPC cells, EBV-miR-BART10-3p directly targets BTRC gene that encodes βTrCP (beta-transducin repeat containing E3 ubiquitin protein ligase). We found that EBV-miR-BART10-3p expression in clinical samples from a cohort of 106 NPC patients negatively correlated with BTRC expression levels. Over-expression of EBV-miR-BART10-3p and down-regulation of BTRC were associated with poor prognosis in NPC patients. EBV-miR-BART10-3p promoted the invasion and migration cabilities of NPC cells through the targeting of BTRC and regulation of the expression of the downstream substrates β-catenin and Snail. As a result, EBV-miR-BART10-3p facilitated epithelial-mesenchymal transition of NPC. Our study presents an unreported mechanism underlying EBV infection in NPC carcinogenesis, and provides a potential novel biomarker for NPC diagnosis, treatment and prognosis.
Sigma-Aldrich
MISSION® esiRNA, targeting human BTRC
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O