Merck
  • Comprehensive characterization of sphingolipid ceramide N-deacylase for the synthesis and fatty acid remodeling of glycosphingolipids.
Comprehensive characterization of sphingolipid ceramide N-deacylase for the synthesis and fatty acid remodeling of glycosphingolipids.
Applied microbiology and biotechnology (2015-02-17)
Yun-Bin Han, Lie Wu, Jamie R Rich, Feng-Tao Huang, Stephen G Withers, Yan Feng, Guang-Yu Yang
ABSTRACT
Sphingolipid ceramide N-deacylase (SCDase) catalyzes reversible reactions in which the amide linkage in glycosphingolipids is hydrolyzed or synthesized. While SCDases show great value for the enzymatic synthesis of glycosphingolipids, they are relatively poorly characterized enzymes. In this work, the enzymatic properties of SCDase from Shewanella alga G8 (SA_SCD) were systematically characterized and compared with the commercially available SCDase from Pseudomonas sp. TK4 (PS_SCD). The optimal pH values for the hydrolytic and synthetic activity of SA_SCD were pH 6.0 and pH 7.5, respectively. Both activities were strongly inhibited by Zn(2+) and Cu(2+), while Fe(2+), Co(2+), Ni(2+), Mn(2+), Ca(2+), and Mg(2+) promoted the hydrolytic activity but inhibited the synthetic activity. SA_SCD showed very broad substrate specificity both in hydrolysis and synthesis. Importantly, SA_SCD has a broader specificity for acyl donor acceptance than does PS_SCD, especially for unsaturated fatty acids and fatty acids with very short or long acyl chains. Further kinetic analysis revealed that the k cat/K M value for the hydrolytic activity of SA_SCD was 8.9-fold higher than that of PS_SCD for GM1a, while the values for the synthetic activity were 38-fold higher for stearic acid and 23-fold higher for lyso-GM1a (d18:1) than those of PS_SCD, respectively. The broad fatty acid specificity and high catalytic efficiency, together with the ease of expression of SA_SCD in Escherichia coli, make it a better biocatalyst than is PS_SCD for the synthesis and structural remodeling of glycosphingolipids.
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Octylphosphonic acid, 97%
Sigma-Aldrich
Phthaldialdehyde, ≥99.0% (HPLC)