Merck
  • CuCl2-catalyzed one-pot formation of tetrahydroquinolines from N-methyl-N-alkylanilines and vinyl ethers in the presence of t-butylhydroperoxide.
CuCl2-catalyzed one-pot formation of tetrahydroquinolines from N-methyl-N-alkylanilines and vinyl ethers in the presence of t-butylhydroperoxide.
Molecules (Basel, Switzerland) (2007-11-17)
Xianghua Yang, Chanjuan Xi, Yanfeng Jiang
ABSTRACT
Tetrahydroquinoline skeletons can be formed by a CuCl2-catalyzed one-pot reaction of N-methyl-N-alkylanilines and vinyl ethers in the presence of t-butyl-hydroperoxide.
Sigma-Aldrich
4,N,N-Trimethylaniline, ≥98.5% (GC)
Sigma-Aldrich
4,N,N-Trimethylaniline, purum, ≥98.0% (GC)