Merck
  • Phosphorylation of ARD1 by IKKbeta contributes to its destabilization and degradation.
Phosphorylation of ARD1 by IKKbeta contributes to its destabilization and degradation.
Biochemical and biophysical research communications (2009-09-01)
Hsu-Ping Kuo, Dung-Fang Lee, Weiya Xia, Chien-Chen Lai, Long-Yuan Li, Mien-Chie Hung
ABSTRACT
IkappaB kinase beta (IKKbeta), a major kinase downstream of various proinflammatory signals, mediates multiple cellular functions through phosphorylation and regulation of its substrates. On the basis of protein sequence analysis, we identified arrest-defective protein 1 (ARD1), a protein involved in apoptosis and cell proliferation processes in many human cancer cells, as a new IKKbeta substrate. We provided evidence showing that ARD1 is indeed a bona fide substrate of IKKbeta. IKKbeta physically associated with ARD1 and phosphorylated it at Ser209. Phosphorylation by IKKbeta destabilized ARD1 and induced its proteasome-mediated degradation. Impaired growth suppression was observed in ARD1 phosphorylation-mimic mutant (S209E)-transfected cells as compared with ARD1 non-phosphorylatable mutant (S209A)-transfected cells. Our findings of molecular interactions between ARD1 and IKKbeta may enable further understanding of the upstream regulation mechanisms of ARD1 and of the diverse functions of IKKbeta.
Sigma-Aldrich
Anti-acetyl-Lysine Antibody, clone 4G12, clone 4G12, Upstate®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-acetyl-Lysine Antibody, serum, Upstate®