Merck
  • A pilot study of the efficacy of a computerized executive functioning remediation training with game elements for children with ADHD in an outpatient setting: outcome on parent- and teacher-rated executive functioning and ADHD behavior.
A pilot study of the efficacy of a computerized executive functioning remediation training with game elements for children with ADHD in an outpatient setting: outcome on parent- and teacher-rated executive functioning and ADHD behavior.
Journal of attention disorders (2012-08-11)
S van der Oord, A J G B Ponsioen, H M Geurts, E L Ten Brink, P J M Prins
ABSTRACT
This pilot study tested the short- and long-term efficacy (9 weeks follow-up) of an executive functioning (EF) remediation training with game elements for children with ADHD in an outpatient clinical setting, using a randomized controlled wait-list design. Furthermore, in a subsample, that is, those treated with methylphenidate, additive effects of the EF training were assessed. A total of 40 children (aged 8-12 years) were randomized to the EF training or wait-list. The training consisted of a 25-session training of inhibition, cognitive flexibility, and working memory. Treatment outcome was assessed by parent- and teacher-rated EF, ADHD, oppositional deviant disorder, and conduct disorder symptoms. Children in the EF training showed significantly more improvement than those in the wait-list condition on parent-rated EF and ADHD behavior in the total sample and in the subsample treated with methylphenidate. Effects were maintained at follow-up. This pilot study shows promising evidence for the efficacy of an EF training with game elements.
Sigma-Aldrich
Methylphenidate hydrochloride
Supelco
Methylphenidate hydrochloride solution, ampule of 1 mL, (Racemic mixture), 1.0 mg/mL in methanol (as free base), certified reference material, Cerilliant®