Merck
  • An updated review of acitretin--a systemic retinoid for the treatment of psoriasis.
An updated review of acitretin--a systemic retinoid for the treatment of psoriasis.
Expert opinion on drug metabolism & toxicology (2008-07-16)
Mei-Lin Pang, Jenny E Murase, John Koo
ABSTRACT
Acitretin is a systemic retinoid used for psoriasis. It normalizes cellular differentiation and maturation and is also used as a chemopreventive agent against cutaneous malignancies. However, it is not used frequently because of its side-effect profile. Safety and efficacy of acitretin was evaluated as monotherapy, as well as in combination with other systemic agents. Medical literature from 2005 to 2008 was reviewed. The most scientifically rigorous clinical trials were selected for Psoriasis Area and Severity Index. Articles were limited to case reports or clinical trials, human subjects and English language journals. Acitretin is effective as monotherapy for pustular and erythrodermic psoriasis and for plaque psoriasis (with other systemic agents). Side effects of acitretin use occur more commonly with high doses. Hence, acitretin is safe and effective for psoriasis.
Sigma-Aldrich
Acitretin, ≥98.0% (HPLC)
Acitretin, European Pharmacopoeia (EP) Reference Standard
USP
Acitretin, United States Pharmacopeia (USP) Reference Standard