Merck
  • Perindopril/indapamide 2/0.625 mg/day: a review of its place in the management of hypertension.
Perindopril/indapamide 2/0.625 mg/day: a review of its place in the management of hypertension.
Drugs (2001-07-24)
A J Matheson, S M Cheer, K L Goa
ABSTRACT
The fixed low-dose combination of the ACE inhibitor perindopril and the non-thiazide diuretic indapamide has been evaluated in the management of patients with mild to moderate hypertension. Combination therapy aims to improve overall therapeutic efficacy while minimising adverse effects. In well-designed multicentre clinical trials, perindopril/indapamide at doses ranging from 2/0.625 to 8/2.5 mg/day was significantly more effective than placebo in achieving adequate blood pressure (BP) control. A similar reduction in supine BP was observed when combined perindopril/indapamide 2/0.625 mg/day was compared with losartan 50 mg/day or atenolol 50 mg/day. Similar reductions in 24-hour ambulatory BP were also seen with perindopril/indapamide 2/0.625 mg/day and irbesartan 150 mg/day. However, response and normalisation rates were significantly higher with combination therapy than with losartan or irbesartan monotherapy. Combined perindopril/indapamide 2/0.625 mg/day therapy effectively reduced BP in elderly patients aged 65 to 85 years to a significantly greater extent than either atenolol 50 mg/day or placebo. Supine BP was also normalised in approximately two-thirds of patients in a small noncomparative trial in patients with hypertension and renal impairment. Low-dose perindopril/indapamide 2/0.625 mg/day was well tolerated in clinical trials; the most common adverse events were headache and cough. Hypokalaemia, associated with the use of diuretics, occurred with a higher incidence with combined perindopril/indapamide 2/0.625 mg/day therapy than with either atenolol 50 mg/day or placebo. Perindopril/indapamide 2/0.625 mg/day has shown efficacy in well designed comparative trials with atenolol, losartan and irbesartan including elderly patients and patients with renal impairment. Studies comparing this dosage of perindopril/ indapamide with other combination therapies would be beneficial in allowing the place of perindopril/indapamide to be more accurately determined. The fixed-low dose combination of perindopril/indapamide provides a promising and well tolerated treatment option in the management of patients with mild to moderate hypertension.
Product Description
Supelco
Indapamide, analytical standard, for drug analysis
Indapamide, European Pharmacopoeia (EP) Reference Standard
USP
Indapamide, United States Pharmacopeia (USP) Reference Standard