Merck
  • Base-controlled selective conversion of Michael adducts of malonates with enones in the presence of iodine.
Base-controlled selective conversion of Michael adducts of malonates with enones in the presence of iodine.
The Journal of organic chemistry (2011-10-27)
Chun-Bao Miao, Min Zhang, Zong-Yong Tian, Hai-Tao Xi, Xiao-Qiang Sun, Hai-Tao Yang
ABSTRACT
An efficient base-controlled selective conversion of the Michael adducts of malonates with enones in the presence of iodine is reported. Highly functionalized cyclopropane, oxetane, and α-hydroxylmalonate derivatives are obtained selectively using DBU, Na(2)CO(3), and NaOAc as the base, respectively. O(2) was identified to be crucial to the formation of oxetane and α-hydroxylmalonate derivatives.
Sigma-Aldrich
Trimethylene oxide, 97%